3',5'-Difluoro-4'-hydroxypropiophenone ≥98%
Supplier: Matrix Scientific
004375-5G
004375-1G
101787-126EA
226.15
USD
101787-126
101787-124
3',5'-Difluoro-4'-hydroxypropiophenone ≥98%
3',5'-Difluoro-4'-hydroxypropiophenone
Hazard code(s): Irritant
Formula:
C₉H₈F₂O₂ |
MDL Number:
MFCD00061287 CAS Number: 178374-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...