Sodium methanesulfinate ≥90%

Supplier: Matrix Scientific

Synonyms: Methanesulfinic acid, sodium saltMethanesulphinic acid, sodium salt

004309-100G
101787-080EA 161.54 USD
101787-080
Sodium methanesulfinate ≥90%
Sodium methanesulphinate

Hazard code(s): Irritant

Formula: CH₃NaO₂S
MW: 102.09 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD00040392
CAS Number: 20277-69-4
EINECS: 243-669-6

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR