2-Fluorobiphenyl ≥99%
Supplier: Matrix Scientific
Synonyms:
PFB No. 1
, 2-Fluoro-1,1'-biphenyl
004215-100G
101786-966EA
192.31
USD
101786-966
2-Fluorobiphenyl ≥99%
2-Fluorobiphenyl
Hazard code(s): Irritant
Formula:
C₁₂H₉F MW: 172.2 g/mol Boiling Pt: 248 °C Melting Pt: 73…74.5 °C Density: 1.2452 g/cm³ (25 °C) |
MDL Number:
MFCD00000317 CAS Number: 321-60-8 EINECS: 206-290-7 |
Specification Test Results
Boiling point | 248°C |
Melting point | 73-74.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...