2-Nitrothiophene tech. 80%

Supplier: Matrix Scientific

004201-25G 004201-100G
101786-936EA 72.31 USD
101786-936 101786-938
2-Nitrothiophene tech. 80%
2-Nitrothiophene

  • RTECS: XN0030000
  • TSCA: T

Formula: C₄H₃NO₂S
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: 46.5 °C
Density: 1.3644 g/cm³ (42.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005425
CAS Number: 609-40-5
EINECS: 210-190-9

Order Now

Specification Test Results

Boiling point 224-225°C
Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR