Dimethyl-2-iodoterephthalate ≥98%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-iodoterephthalate

004194-25G 004194-5G
101786-928EA 144.9 USD
101786-928 101786-930
Dimethyl-2-iodoterephthalate ≥98%
Dimethyl 2-iodoterephthalate

Hazard code(s): Irritant

Formula: C₁₀H₉IO₄
MW: 320.08 g/mol
Melting Pt: 81…82 °C
MDL Number: MFCD00060640
CAS Number: 165534-79-2

Order Now

Specification Test Results

Melting point 81-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR