2-Chlorocinnamic acid ≥99%

Supplier: Matrix Scientific
Warning

004176-100G
101786-882EA 58.46 USD
101786-882
2-Chlorocinnamic acid ≥99%
2-Chlorocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₇ClO₂
MW: 182.61 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 208…210 °C
MDL Number: MFCD00004372
CAS Number: 3752-25-8
EINECS: 223-154-2

Order Now

Specification Test Results

Melting point 208-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR