3-Iodopyridine ≥98%

Supplier: Matrix Scientific
Warning

004119-5G 004119-25G
101786-852EA 21.54 USD
101786-852 101786-854
3-Iodopyridine ≥98%
3-Iodopyridine

Hazard code(s): Irritant, Light Sensitive

Formula: C₅H₄IN
MW: 205 g/mol
Boiling Pt: 90…92 °C (14 mmHg)
Melting Pt: 53…56 °C
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now

Specification Test Results

Melting point 53-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR