2,6-Dimethylthiophenol ≥97%
Supplier: Matrix Scientific
|
Warning
|
004100-100G
004100-25G
101786-832EA
140
USD
101786-832
101786-834
2,6-Dimethylthiophenol ≥97%
2,6-Dimethylthiophenol
Hazard code(s): Toxic, Stench
Formula:
C₈H₁₀S MW: 138.23 g/mol Boiling Pt: 122 °C (50mm) Density: 1.038 g/cm³ |
MDL Number:
MFCD00010021 CAS Number: 118-72-9 EINECS: 204-272-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 122°C/50mm |
Density | 1.038 |
ND20 | 1.5749 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...