4-Chloromethyl-o-xylene ≥98%

Supplier: Matrix Scientific

Synonyms: α-Chloro-1,3,4-trimethylbenzene1-(Chloromethyl)-3,4-dimethylbenzene4-(Chloromethyl)-1,2-dimethylbenzene3,4-Dimethylbenzyl chloride

004069-25G
101786-778EA 101.2 USD
101786-778
4-Chloromethyl-o-xylene ≥98%
4-Chloromethyl-o-xylene
Formula: C₉H₁₁Cl
MW: 154.64 g/mol
MDL Number: MFCD00000910
CAS Number: 102-46-5
EINECS: 203-032-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 116-117°C/2
Density 1.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR