2,3-Dimethylbenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
004060-100G
004060-25G
101786-744EA
41.54
USD
101786-744
101786-746
2,3-Dimethylbenzoic acid ≥98%
2,3-Dimethylbenzoic acid
Hazard code(s): Irritant
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 282 °C (1013 hPa) Melting Pt: 145…147 °C Density: 1.283 g/cm³ (20 °C) |
MDL Number:
MFCD00002479 CAS Number: 603-79-2 EINECS: 210-058-0 |
Specification Test Results
Melting point | 145-147°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...