5-Methyl-2-thiopheneboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Methylthiophene-2-boronic acid(5-Methylthiophen-2-yl)boronic acid

003968-5G
101786-542EA 101.54 USD
101786-542
5-Methyl-2-thiopheneboronic acid ≥97%
5-Methyl-2-thiopheneboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₇BO₂S
MW: 141.99 g/mol
Melting Pt: ∼160 °C (dec.)
MDL Number: MFCD01318166
CAS Number: 162607-20-7
ADR: lorem ipsum,lorem ipsum

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR