Benzo[b]thiophene-2-boronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Benzothienylboronic acid
, Benzo[b]thiophen-2-ylboronic acid
003964-1G
101786-528EA
13.85
USD
101786-528
Benzo[b]thiophene-2-boronic acid ≥97%
Benzo[b]thiophene-2-boronic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₈H₇BO₂S MW: 178.02 g/mol Melting Pt: 255…257 °C |
MDL Number:
MFCD01075674 CAS Number: 98437-23-1 |
Specification Test Results
Melting point | 255-257°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...