Perfluorohexane ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: Tetradecafluorohexane

003943-25G 003943-100G
102832-758EA 41.54 USD
102832-758 101786-486
Perfluorohexane ≥98%
Perfluorohexane

Hazard code(s): Irritant, Refrigerate

Formula: C₆F₁₄
MW: 338.04 g/mol
Boiling Pt: 58…60 °C
Melting Pt: –4 °C
Density: 1.669 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000437
CAS Number: 355-42-0
EINECS: 206-585-0

Order Now

Specification Test Results

Boiling point 58-60°C
Density 1.669
ND20 1.252

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR