4-Acetylphenylboronic acid ≥98%
Supplier: Matrix Scientific
003911-5G
003911-25G
101786-428EA
47.69
USD
101786-428
101786-430
4-Acetylphenylboronic acid ≥98%
4-Acetylphenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₈H₉BO₃ MW: 163.97 g/mol Melting Pt: 258 °C (dec.) |
MDL Number:
MFCD01074667 CAS Number: 149104-90-5 |
Specification Test Results
Melting point | 258°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...