2-Fluorophenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (2-Fluorophenyl)metaboric acid(2-Fluorophenyl)boronic acid2-Fluorobenzeneboronic acidDihydroxy(2-fluorophenyl)borane

003908-100G
102832-756EA 196.92 USD
102832-756
2-Fluorophenylboronic acid ≥98%
2-Fluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Melting Pt: 101…110 °C
MDL Number: MFCD00674013
CAS Number: 1993-03-9

Order Now

Specification Test Results

Melting point 101-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR