(3-Fluorophenyl)acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(3-Fluorophenyl)acetic acid

003892-100G
101786-380EA 64.62 USD
101786-380
(3-Fluorophenyl)acetic acid ≥98%
(3-Fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 151 °C (0.8mm)
Melting Pt: 42…44 °C
MDL Number: MFCD00004331
CAS Number: 331-25-9
EINECS: 206-360-7

Order Now

Specification Test Results

Boiling point 151°C/0.8mm
Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR