Phenylsulfonylacetone ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Propanoylsulfonyl)benzol(2-Propanoylsulphonyl)benzol1-(Phenylsulfonyl)acetone1-(Phenylsulfonyl)propan-2-one1-(Phenylsulphonyl)acetone1-(Phenylsulphonyl)propan-2-oneBenzenesulphonylacetoneBenzenesulfonylacetone

003838-1G
101786-302EA 29.23 USD
101786-302
Phenylsulfonylacetone ≥97%
Phenylsulphonylacetone
Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
MDL Number: MFCD00047794
CAS Number: 5000-44-2

Order Now

Specification Test Results

Melting point 54-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR