2,4,6-Trifluorophenol ≥99%
Supplier: Matrix Scientific
003778-5G
101786-216EA
95.38
USD
101786-216
2,4,6-Trifluorophenol ≥99%
2,4,6-Trifluorophenol
Hazard code(s): Irritant-Harmful
Formula:
C₆H₃F₃O MW: 148.08 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: 49…51 °C |
MDL Number:
MFCD00061216 CAS Number: 2268-17-9 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 49-51°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...