2,3,6-Trifluorophenol ≥98%
Supplier: Matrix Scientific
003777-5G
003777-1G
101786-210EA
27.69
USD
101786-210
101786-212
2,3,6-Trifluorophenol ≥98%
2,3,6-Trifluorophenol
Hazard code(s): Irritant
Formula:
C₆H₃F₃O MW: 148.08 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: 33…37 °C |
MDL Number:
MFCD00061214 CAS Number: 113798-74-6 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 33-37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...