Pentafluoroiodobenzene ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Iodopentafluorobenzene
003673-100G
101786-060EA
740
USD
101786-060
Pentafluoroiodobenzene ≥99%
Pentafluoroiodobenzene
Hazard code(s): Irritant
Formula:
C₆F₅I MW: 293.96 g/mol Boiling Pt: 166 °C Melting Pt: –29 °C Density: 2.204 g/cm³ |
MDL Number:
MFCD00001032 CAS Number: 827-15-6 EINECS: 212-565-2 |
Specification Test Results
Boiling point | 166°C |
Density | 2.204 |
ND20 | 1.496 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...