2,3,6-Trifluorobenzamide ≥97%
Supplier: Matrix Scientific
003664-5G
003664-1G
101786-042EA
38.46
USD
101786-042
101786-044
2,3,6-Trifluorobenzamide ≥97%
2,3,6-Trifluorobenzamide
Hazard code(s): Irritant
Formula:
C₇H₄F₃NO Melting Pt: 215…217 °C |
MDL Number:
MFCD00061266 CAS Number: 207986-22-9 |
Specification Test Results
Melting point | 215-217°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...