3,4,5-Trifluorobenzylamine

Supplier: Matrix Scientific
Danger

Synonyms: (3,4,5-Trifluorophenyl)methanamine

003661-1G
101786-040EA 60 USD
101786-040
3,4,5-Trifluorobenzylamine
3,4,5-Trifluorobenzylamine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 169 °C (1013 hPa)
Melting Pt: 44 °C
MDL Number: MFCD00236318
CAS Number: 235088-69-4
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR