2,4,5-Trifluorobenzylamine

Supplier: Matrix Scientific
Danger

003660-1G
101786-038EA 78.46 USD
101786-038
2,4,5-Trifluorobenzylamine
2,4,5-Trifluorobenzylamine

Hazard code(s): Corrosive, Air Sensitive

Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 55 °C (1mm)
MDL Number: MFCD00236316
CAS Number: 168644-93-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR