2,3,4-Trifluorobenzamide ≥97%

Supplier: Matrix Scientific

003631-1G 003631-5G
101785-968EA 43.08 USD
101785-968 101785-970
2,3,4-Trifluorobenzamide ≥97%
2,3,4-Trifluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₄F₃NO
Melting Pt: 127…129 °C
MDL Number: MFCD00061265
CAS Number: 207919-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR