3,4,5-Trifluorobenzaldehyde ≥98%

Supplier: Matrix Scientific

003629-25G 003629-100G 003629-5G
101785-958EA 38.46 USD
101785-958 101785-962 101785-960
3,4,5-Trifluorobenzaldehyde ≥98%
3,4,5-Trifluorobenzaldehyde

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 174 °C
Melting Pt: 24…26 °C
Density: 1.42 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00083523
CAS Number: 132123-54-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR