2,3,4-Trifluoroanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2,3-Trifluoro-4-methoxybenzene

003618-25G 003618-1G 003618-5G
101785-936EA 461.54 USD
101785-936 101785-934 101785-932
2,3,4-Trifluoroanisole ≥97%
2,3,4-Trifluoroanisole

Hazard code(s): Irritant

Formula: C₇H₅F₃O
MW: 162.11 g/mol
MDL Number: MFCD00042396
CAS Number: 203245-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR