2,4,6-Trifluoroaniline ≥97%

Supplier: Matrix Scientific

003615-25G
101785-926EA 267.69 USD
101785-926
2,4,6-Trifluoroaniline ≥97%
2,4,6-Trifluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 57 °C (22mm)
Melting Pt: 33…34 °C
MDL Number: MFCD00007650
CAS Number: 363-81-5
EINECS: 206-660-8
UN: 2926
ADR: 4.1,III

Order Now

Specification Test Results

Boiling point 57°C/22mm
Melting point 33-34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR