Diethyl hexafluoroglutarate ≥97%
Supplier: Matrix Scientific
Synonyms:
Diethyl perfluoroglutarate
003523-5G
003523-1G
101785-838EA
200
USD
101785-838
101785-836
Diethyl hexafluoroglutarate ≥97%
Diethyl hexafluoroglutarate
Hazard code(s): Irritant
Formula:
C₉H₁₀F₆O₄ MW: 296.17 g/mol Boiling Pt: 75…80 °C (3mm) Density: 1.344 g/cm³ |
MDL Number:
MFCD00015156 CAS Number: 424-40-8 EINECS: 207-035-2 |
Specification Test Results
Boiling point | 75-80°C/3mm |
Density | 1.344 |
ND20 | 1.359 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...