1-Nitro-2,3,4,5-tetrafluorobenzene ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2,3,4,5-Tetrafluoronitrobenzene
, 1,2,3,4-Tetrafluoro-5-nitrobenzene
003503-25G
003503-5G
003503-100G
101785-806EA
269.23
USD
101785-806
101785-804
101785-802
1-Nitro-2,3,4,5-tetrafluorobenzene ≥99%
1-Nitro-2,3,4,5-tetrafluorobenzene
Hazard code(s): Irritant
Formula:
C₆HF₄NO₂ MW: 195.07 g/mol Boiling Pt: 80 °C (24mm) Density: 1.618 g/cm³ |
MDL Number:
MFCD00014686 CAS Number: 5580-79-0 EINECS: 226-972-8 |
Specification Test Results
Boiling point | 80°C/24mm |
Density | 1.618 |
ND20 | 1.473 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...