Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,5,6-Tetrafluoroaniline ≥98%

Supplier: Matrix Scientific

Warning

003487-100G 003487-25G
101785-780EA 258.92 USD InStock
101785-780 101785-778
2,3,5,6-Tetrafluoroaniline ≥98%
2,3,5,6-Tetrafluoroaniline
Hazard code(s): Irritant, Refrigerate


Formula: C₆H₃F₄N
MW: 165.09 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 31 to 32 °C
Density: 1.744 g/cm³ (20 °C)
MDL Number: MFCD00007644
CAS Number: 700-17-4
EINECS: 211-840-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 158°C
Melting point 31-32°C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty