p-Tolylacetic acid ≥99%
Supplier: Matrix Scientific
Synonyms:
(4-Methylphenyl)acetic acid
, 4-Tolylacetic acid
, 2-(p-Tolyl)acetic acid
003421-25G
003421-100G
101785-684EA
70.77
USD
101785-684
101785-682
p-Tolylacetic acid ≥99%
p-Tolylacetic acid
Hazard code(s): Irritant
Formula:
(C₆H₄(CH₃))CH₂COOH MW: 150.18 g/mol Boiling Pt: 265…267 °C Melting Pt: 90…93 °C |
MDL Number:
MFCD00004353 CAS Number: 622-47-9 EINECS: 210-738-7 |
Specification Test Results
Boiling point | 265-267°C |
Melting point | 90-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...