Phenoxyacetamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Phenoxyacetamide

003367-25G 003367-5G
101785-622EA 90.77 USD
101785-622 101785-624
Phenoxyacetamide ≥97%
Phenoxyacetamide

Hazard code(s): Irritant

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD00017140
CAS Number: 621-88-5
EINECS: 210-713-0

Order Now

Specification Test Results

Melting point 101-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR