α,α,α-Trifluoro-p-xylene

Supplier: Matrix Scientific
Danger

Synonyms: 1-Methyl-4-(trifluoromethyl)benzene4-Methylbenzotrifluoride4-(Trifluoromethyl)toulene

003309-5G
101785-568EA 60 USD
101785-568
α,α,α-Trifluoro-p-xylene
α,α,α-Trifluoro-p-xylene

Hazard code(s): Flammable

Formula: C₈H₇F₃
MW: 160.14 g/mol
Boiling Pt: 129 °C
Density: 1.144 g/cm³
MDL Number: MFCD00075476
CAS Number: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 129°C
Density 1.144
ND20 1.426

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR