3-Fluoro-α,α,α-trichlorotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Fluorobenzotrichloride
, m-Fluoro-α,α,α-trichlorotoluene
003304-1G
101785-564EA
55.38
USD
101785-564
3-Fluoro-α,α,α-trichlorotoluene ≥98%
3-Fluoro-α,α,α-trichlorotoluene
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₄Cl₃F MW: 213.47 g/mol Boiling Pt: 119.4 °C (45 mmHg) Density: 1.4408 g/cm³ (30 °C) |
MDL Number:
MFCD00042073 CAS Number: 401-77-4 EINECS: 206-931-0 |
Specification Test Results
ND20 | 1.537 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...