2-Fluorobenzothiazole ≥99%

Supplier: Matrix Scientific

003302-1G
101785-562EA 81.54 USD
101785-562
2-Fluorobenzothiazole ≥99%
2-Fluorobenzothiazole

Hazard code(s): Irritant

Formula: C₇H₄FNS
MW: 153.18 g/mol
Density: 1.33 g/cm³
MDL Number: MFCD00047254
CAS Number: 1123-98-4
EINECS: 214-384-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.33

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR