2-Fluorobenzonitrile ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
o-Fluorobenzonitrile
003299-100G
003299-500G
101785-552EA
223.08
USD
101785-552
101785-550
2-Fluorobenzonitrile ≥99%
2-Fluorobenzonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₄FN MW: 121.11 g/mol Boiling Pt: 90 °C (21mm) Melting Pt: 35…37 °C Density: 1.116 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00001773 CAS Number: 394-47-8 EINECS: 206-897-7 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 90°C/21mm |
Density | 1.116 |
ND20 | 1.5083 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...