2-Fluorobenzoic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
o-Fluorobenzoic acid
003291-2.5KG
003291-100G
101785-528EA
316.48
USD
101785-528
101785-524
2-Fluorobenzoic acid ≥97%
2-Fluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₅FO₂ MW: 140.11 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 123…125 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |
Specification Test Results
Melting point | 123-125°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...