4-Fluorobenzamide ≥98%

Supplier: Matrix Scientific

003263-100G
101785-470EA 60 USD
101785-470
4-Fluorobenzamide ≥98%
4-Fluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₆FNO
MW: 139.13 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 154…156 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00007992
CAS Number: 824-75-9
EINECS: 212-535-9

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR