4-Fluorobenzaldehyde ≥98%
Supplier: Matrix Scientific
003257-500G
003257-100G
101785-460EA
169.23
USD
101785-460
101785-458
4-Fluorobenzaldehyde ≥98%
4-Fluorobenzaldehyde
Hazard code(s): Flammable
Formula:
C₇H₅FO MW: 124.11 g/mol Boiling Pt: 176…179 °C Melting Pt: –10 °C Density: 1.157 g/cm³ Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003378 CAS Number: 459-57-4 EINECS: 207-293-6 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 176-179°C |
Melting point | -10°C |
Density | 1.157 |
ND20 | 1.52 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...