3-Ethylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

003211-5G
101785-412EA 209.23 USD
101785-412
3-Ethylphenyl isothiocyanate ≥98%
3-Ethylphenyl isothiocyanate

Hazard code(s): Toxic, Lachrymator, Refrigerate

Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 115…116 °C (7mm)
Density: 1.07 g/cm³
MDL Number: MFCD00041084
CAS Number: 19241-20-4
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 115-116°C/7
Density 1.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR