4-Ethylphenethylamine ≥98%

Supplier: Matrix Scientific

003184-1G 003184-5G
101785-376EA 110.77 USD
101785-376 101785-378
4-Ethylphenethylamine ≥98%
4-Ethylphenethylamine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₁₀H₁₅N
Boiling Pt: 70 °C (1mm)
Density: 0.93 g/cm³
MDL Number: MFCD00079757
CAS Number: 64353-29-3

Order Now

Specification Test Results

Boiling point 70°C/1mm
Density 0.93

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR