4-Bromophenethylamine hydrobromide

Supplier: Matrix Scientific

003183-5G 003183-1G
101785-372EA 144.9 USD
101785-372 101785-374
4-Bromophenethylamine hydrobromide
4-Bromophenethylamine hydrobromide

Hazard code(s): Irritant

Formula: C₈H₁₁Br₂N
Melting Pt: 272…274 °C
MDL Number: MFCD00060723
CAS Number: 206559-45-7

Order Now

Specification Test Results

Melting point 272-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR