3-Tolualdehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
m-Tolylaldehyde
, m-Tolualdehyde
, 3-Methylbenzaldehyde
003174-25G
003174-100G
101785-358EA
52.31
USD
101785-358
101785-356
3-Tolualdehyde ≥95%
3-Tolualdehyde
Hazard code(s): Air Sensitive
Formula:
C₈H₈O MW: 120.15 g/mol Boiling Pt: 199 °C Density: 1.019 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003374 CAS Number: 620-23-5 EINECS: 210-632-0 |
Specification Test Results
Boiling point | 199°C |
Density | 1.019 |
ND20 | 1.541 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...