Methyl indole-3-carboxylate ≥99%
Supplier: Matrix Scientific
003154-100G
003154-25G
101785-328EA
198.46
USD
101785-328
101785-326
Methyl indole-3-carboxylate ≥99%
Methyl indole-3-carboxylate
Hazard code(s): Irritant, Store in Freezer
Formula:
C₁₀H₉NO₂ Melting Pt: 149…152 °C |
MDL Number:
MFCD00189407 CAS Number: 942-24-5 |
Specification Test Results
Melting point | 149-152°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...