1-Cyanocyclopropanecarboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Cyano-1-cyclopropanecarboxylic acid

003112-25G 003112-5G
101785-262EA 201.25 USD
101785-262 101785-260
1-Cyanocyclopropanecarboxylic acid ≥98%
1-Cyanocyclopropanecarboxylic acid

Hazard code(s): Toxic, Corrosive

Formula: C₅H₅NO₂
MW: 111.1 g/mol
Melting Pt: 143…149 °C
MDL Number: MFCD00190651
CAS Number: 6914-79-0
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Boiling point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR