Dimethyl-2-chloroterephthalate ≥98%

Supplier: Matrix Scientific

003103-25G 003103-5G
101785-248EA 623.08 USD
101785-248 101785-246
Dimethyl-2-chloroterephthalate ≥98%
Dimethyl 2-chloroterephthalate

Hazard code(s): Irritant

Formula: C₁₀H₉ClO₄
Melting Pt: 58…60 °C
MDL Number: MFCD00060639
CAS Number: 18643-84-0
EINECS: 242-475-9

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR