2,3,5-Trichlorobenzoic acid ≥95%
Supplier: Matrix Scientific
|
Warning
|
003091-5G
003091-1G
101785-214EA
131.01
USD
101785-214
101785-212
2,3,5-Trichlorobenzoic acid ≥95%
2,3,5-Trichlorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₃Cl₃O₂ MW: 225.46 g/mol Boiling Pt: 324 °C (1013 hPa) Melting Pt: 164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060674 CAS Number: 50-73-7 |
Specification Test Results
Melting point | 164°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...