Ethyl isothiocyanatoformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethoxycarbonyl isothiocyanate
003034-25G
003034-5G
101785-116EA
83.95
USD
101785-116
101785-114
Ethyl isothiocyanatoformate ≥97%
Ethyl isothiocyanatoformate
Hazard code(s): Flammable, Corrosive, Moisture Sensitive, Refrigerate
Formula:
C₄H₅NO₂S MW: 131.16 g/mol Boiling Pt: 56 °C (18mm) Density: 1.112 g/cm³ |
MDL Number:
MFCD00004814 CAS Number: 16182-04-0 EINECS: 240-318-9 |
Specification Test Results
Boiling point | 56°C/18mm |
Density | 1.112 |
ND20 | 1.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...