1-Fluoronaphthalene ≥99%

Supplier: Matrix Scientific
Warning

002984-100G
101785-002EA 101.2 USD
101785-002
1-Fluoronaphthalene ≥99%
1-Fluoronaphthalene

Hazard code(s): Irritant

Formula: C₁₀H₇F
MW: 146.16 g/mol
Boiling Pt: 215 °C
Melting Pt: –13 °C
Density: 1.332 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003873
CAS Number: 321-38-0
EINECS: 206-287-0

Order Now

Specification Test Results

Boiling point 215°C
Melting point -13°C
Density 1.332
ND20 1.593

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR