1H,1H,2H-Perfluoro-1-decene ≥97%
Supplier: Matrix Scientific
|
Warning
|
002863-100G
002863-5G
002863-25G
101784-792EA
143.08
USD
101784-792
101784-790
101784-788
1H,1H,2H-Perfluoro-1-decene ≥97%
1H,1H,2H-Perfluoro-1-decene
Hazard code(s): Irritant
Formula:
C₁₀H₃F₁₇ MW: 446.11 g/mol Boiling Pt: 146…147 °C Density: 1.677 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00039246 CAS Number: 21652-58-4 EINECS: 244-503-5 |
Specification Test Results
Boiling point | 146-147°C |
Density | 1.677 |
ND20 | 1.3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...